Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHLPP2
Entrez Gene ID
23035
Gene Name
PH domain and leucine rich repeat protein phosphatase 2
Gene Aliases
PHLPPL, PPM3B
Chromosome Location
Chr.16: 71644924 - 71724701 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289003.1 NP_001275932.1 14 - 15 2808 72
NM_015020.3 NP_055835.2 15 - 16 2662 72
GenBank mRNA AB023148.2 - 14 - 15 2808 72
AK001854.1 - 3 - 4 371 72
BC035267.1 - 10 - 11 1497 72
BC129927.1 - 14 - 15 2289 72
BX647823.1 - 15 - 16 2663 72
BX648157.1 - 9 - 10 1398 72

View Full Product Details