The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014003.3 | NP_054722.2 | 6 - 7 | 1236 | 73 |
XM_005256269.1 | XP_005256326.1 | 5 - 6 | 1058 | 73 | |
XM_011523484.1 | XP_011521786.1 | 6 - 7 | 1163 | 73 | |
XM_011523485.1 | XP_011521787.1 | 6 - 7 | 1125 | 73 | |
XM_017023913.1 | XP_016879402.1 | 5 - 6 | 1058 | 73 | |
GenBank mRNA | AF038391.1 | - | 5 - 6 | 888 | 73 |
AK291634.1 | - | 6 - 7 | 1028 | 73 | |
AK293776.1 | - | 7 - 8 | 1007 | 73 | |
AK297269.1 | - | 6 - 7 | 631 | 73 | |
AK309391.1 | - | 4 - 5 | 450 | 73 | |
AK311169.1 | - | 7 - 8 | 808 | 73 | |
BC004235.2 | - | 6 - 7 | 1012 | 73 | |
BC008340.1 | - | 6 - 7 | 1012 | 73 | |
D86977.1 | - | 6 - 7 | 1017 | 73 |