Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CBX7
Entrez Gene ID
23492
Gene Name
chromobox 7
Gene Aliases
-
Chromosome Location
Chr.22: 39130772 - 39152661 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175709.3 NP_783640.1 5 - 6 689 92
XM_005261413.4 XP_005261470.1 5 - 6 531 92
XM_006724174.3 XP_006724237.1 5 - 6 835 92
XM_006724176.3 XP_006724239.1 5 - 6 660 92
XM_006724177.3 XP_006724240.1 5 - 6 633 92
XM_006724178.3 XP_006724241.1 5 - 6 558 92
GenBank mRNA BC051773.1 - 5 - 6 640 92
DQ064603.1 - 5 - 6 321 92

View Full Product Details