The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032853.3 | NP_116242.2 | 9 - 10 | 1538 | 120 |
NR_024247.1 | 9 - 10 | 1607 | 120 | ||
XM_011528380.2 | XP_011526682.1 | 9 - 10 | 1528 | 120 | |
XM_011528382.2 | XP_011526684.1 | 9 - 10 | 1528 | 120 | |
XM_011528383.1 | XP_011526685.1 | 9 - 10 | 1528 | 120 | |
XM_011528384.2 | XP_011526686.1 | 9 - 10 | 1528 | 120 | |
XM_011528385.2 | XP_011526687.1 | 10 - 11 | 1729 | 120 | |
XM_017027401.1 | XP_016882890.1 | 9 - 10 | 1528 | 120 | |
GenBank mRNA | AK075241.1 | - | 3 - 4 | 168 | 120 |
AK309092.1 | - | 8 - 9 | 1426 | 120 | |
AK310721.1 | - | 10 - 11 | 1641 | 120 | |
BC110874.1 | - | 8 - 9 | 1504 | 120 | |
BC130443.1 | - | 9 - 10 | 1567 | 120 | |
BC144138.1 | - | 8 - 9 | 1454 | 120 | |
BC144147.1 | - | 9 - 10 | 1567 | 120 | |
BQ071619.1 | - | 5 - 6 | 569 | 120 |