The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005883.2 | NP_005874.1 | 8 - 9 | 1026 | 88 |
XM_005259475.2 | XP_005259532.1 | 8 - 9 | 910 | 88 | |
XM_006722607.2 | XP_006722670.1 | 8 - 9 | 907 | 88 | |
XM_006722608.3 | XP_006722671.2 | 8 - 9 | 1171 | 88 | |
XM_006722609.3 | XP_006722672.1 | 8 - 9 | 980 | 88 | |
XM_006722610.3 | XP_006722673.2 | 8 - 9 | 1167 | 88 | |
GenBank mRNA | AB012162.1 | - | 7 - 8 | 2531 | 88 |
AF128222.1 | - | 7 - 8 | 1019 | 88 | |
AJ012652.1 | - | 7 - 8 | 1019 | 88 | |
BC020381.1 | - | 8 - 9 | 877 | 88 | |
BC032573.1 | - | 8 - 9 | 1026 | 88 |