Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABCC5
Entrez Gene ID
10057
Gene Name
ATP binding cassette subfamily C member 5
Gene Aliases
ABC33, EST277145, MOAT-C, MOATC, MRP5, SMRP, pABC11
Chromosome Location
Chr.3: 183919934 - 184018010 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320032.1 NP_001306961.1 11 - 12 2058 103
NM_005688.3 NP_005679.2 11 - 12 1943 103
XM_005247058.4 XP_005247115.1 11 - 12 3511 103
XM_005247059.4 XP_005247116.1 11 - 12 1933 103
XM_011512314.1 XP_011510616.1 11 - 12 1834 103
XM_011512315.1 XP_011510617.1 11 - 12 1993 103
XM_017005492.1 XP_016860981.1 8 - 9 3182 103
GenBank mRNA AB005659.1 - 5 - 6 1020 103
AB019002.1 - 10 - 11 1809 103
AB209103.1 - 8 - 9 1453 103
AB209454.1 - 11 - 12 1870 103
AF104942.1 - 11 - 12 1882 103
AY196484.1 - 10 - 11 1769 103
BC140771.1 - 10 - 11 1819 103
U83661.2 - 11 - 12 1876 103

View Full Product Details