Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
USP13
Entrez Gene ID
8975
Gene Name
ubiquitin specific peptidase 13 (isopeptidase T-3)
Gene Aliases
ISOT3, IsoT-3
Chromosome Location
Chr.3: 179653145 - 179789401 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003940.2 NP_003931.2 7 - 8 980 70
XM_011513269.1 XP_011511571.1 7 - 8 927 70
XM_017007425.1 XP_016862914.1 7 - 8 824 70
XM_017007426.1 XP_016862915.1 6 - 7 791 70
XM_017007427.1 XP_016862916.1 4 - 5 511 70
GenBank mRNA AK302404.1 - 7 - 8 789 70
BC016146.1 - 7 - 8 933 70
HM138083.1 - 7 - 8 899 70
U75362.1 - 7 - 8 993 70

View Full Product Details