The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015046.5 | NP_055861.3 | 22 - 23 | 7107 | 165 |
XM_005272171.1 | XP_005272228.1 | 22 - 23 | 7108 | 165 | |
XM_005272172.2 | XP_005272229.1 | 23 - 24 | 7206 | 165 | |
XM_005272173.2 | XP_005272230.1 | 22 - 23 | 7244 | 165 | |
XM_011518404.2 | XP_011516706.1 | 23 - 24 | 7248 | 165 | |
XM_011518405.2 | XP_011516707.1 | 22 - 23 | 7266 | 165 | |
XM_011518406.2 | XP_011516708.1 | 22 - 23 | 7108 | 165 | |
XM_011518407.1 | XP_011516709.1 | 22 - 23 | 7108 | 165 | |
XM_017014495.1 | XP_016869984.1 | 22 - 23 | 7179 | 165 | |
XM_017014496.1 | XP_016869985.1 | 13 - 14 | 1775 | 165 | |
XM_017014497.1 | XP_016869986.1 | 13 - 14 | 2144 | 165 | |
GenBank mRNA | AB014525.2 | - | 20 - 21 | 6884 | 165 |
AK001456.1 | - | 12 - 13 | 1643 | 165 | |
AK025601.1 | - | 7 - 8 | 738 | 165 | |
AY362728.1 | - | 20 - 21 | 6931 | 165 | |
BC032600.2 | - | 13 - 14 | 1684 | 165 | |
BC032622.1 | - | 13 - 14 | 1985 | 165 | |
BC078166.1 | - | 12 - 13 | 1644 | 165 | |
BC106017.1 | - | 11 - 12 | 1495 | 165 | |
BC137350.1 | - | 21 - 22 | 7041 | 165 | |
BX537849.1 | - | 22 - 23 | 7092 | 165 | |
BX538166.1 | - | 22 - 23 | 7110 | 165 | |
CR749249.1 | - | 13 - 14 | 5588 | 165 | |
KU178559.1 | - | 10 - 11 | 1377 | 165 | |
KU178560.1 | - | 10 - 11 | 1377 | 165 |