The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005272171.1 | XP_005272228.1 | 25 - 26 | 7474 | 69 |
XM_005272172.2 | XP_005272229.1 | 26 - 27 | 7572 | 69 | |
XM_005272173.2 | XP_005272230.1 | 25 - 26 | 7610 | 69 | |
XM_011518404.2 | XP_011516706.1 | 26 - 27 | 7614 | 69 | |
XM_011518405.2 | XP_011516707.1 | 25 - 26 | 7632 | 69 | |
XM_017014496.1 | XP_016869985.1 | 16 - 17 | 2141 | 69 | |
GenBank mRNA | AK226007.1 | - | 16 - 17 | 2059 | 69 |
AK302394.1 | - | 3 - 4 | 247 | 69 |