The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005272171.1 | XP_005272228.1 | 26 - 27 | 7556 | 80 |
XM_005272172.2 | XP_005272229.1 | 27 - 28 | 7654 | 80 | |
XM_005272173.2 | XP_005272230.1 | 26 - 27 | 7692 | 80 | |
XM_011518404.2 | XP_011516706.1 | 27 - 28 | 7696 | 80 | |
XM_011518405.2 | XP_011516707.1 | 26 - 27 | 7714 | 80 | |
XM_017014496.1 | XP_016869985.1 | 17 - 18 | 2223 | 80 | |
GenBank mRNA | AK226007.1 | - | 17 - 18 | 2141 | 80 |
AK302394.1 | - | 4 - 5 | 329 | 80 |