The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015046.5 | NP_055861.3 | 10 - 11 | 5457 | 153 |
XM_005272171.1 | XP_005272228.1 | 10 - 11 | 5458 | 153 | |
XM_005272172.2 | XP_005272229.1 | 11 - 12 | 5556 | 153 | |
XM_005272173.2 | XP_005272230.1 | 10 - 11 | 5594 | 153 | |
XM_011518404.2 | XP_011516706.1 | 11 - 12 | 5598 | 153 | |
XM_011518405.2 | XP_011516707.1 | 10 - 11 | 5616 | 153 | |
XM_011518406.2 | XP_011516708.1 | 10 - 11 | 5458 | 153 | |
XM_011518407.1 | XP_011516709.1 | 10 - 11 | 5458 | 153 | |
XM_011518408.2 | XP_011516710.1 | 10 - 11 | 5458 | 153 | |
XM_017014495.1 | XP_016869984.1 | 10 - 11 | 5529 | 153 | |
GenBank mRNA | AB014525.2 | - | 8 - 9 | 5234 | 153 |
AY362728.1 | - | 8 - 9 | 5281 | 153 | |
BC032622.1 | - | 1 - 2 | 335 | 153 | |
BC137350.1 | - | 9 - 10 | 5391 | 153 | |
BX537849.1 | - | 10 - 11 | 5442 | 153 | |
BX538166.1 | - | 10 - 11 | 5460 | 153 | |
CR749249.1 | - | 1 - 2 | 3938 | 153 |