The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015046.5 | NP_055861.3 | 18 - 19 | 6574 | 97 |
XM_005272171.1 | XP_005272228.1 | 18 - 19 | 6575 | 97 | |
XM_005272172.2 | XP_005272229.1 | 19 - 20 | 6673 | 97 | |
XM_005272173.2 | XP_005272230.1 | 18 - 19 | 6711 | 97 | |
XM_011518404.2 | XP_011516706.1 | 19 - 20 | 6715 | 97 | |
XM_011518405.2 | XP_011516707.1 | 18 - 19 | 6733 | 97 | |
XM_011518406.2 | XP_011516708.1 | 18 - 19 | 6575 | 97 | |
XM_011518407.1 | XP_011516709.1 | 18 - 19 | 6575 | 97 | |
XM_011518408.2 | XP_011516710.1 | 18 - 19 | 6575 | 97 | |
XM_017014495.1 | XP_016869984.1 | 18 - 19 | 6646 | 97 | |
XM_017014496.1 | XP_016869985.1 | 9 - 10 | 1242 | 97 | |
XM_017014497.1 | XP_016869986.1 | 9 - 10 | 1611 | 97 | |
GenBank mRNA | AB014525.2 | - | 16 - 17 | 6351 | 97 |
AK001456.1 | - | 8 - 9 | 1110 | 97 | |
AK025601.1 | - | 3 - 4 | 205 | 97 | |
AK226007.1 | - | 9 - 10 | 1160 | 97 | |
AY362728.1 | - | 16 - 17 | 6398 | 97 | |
BC032600.2 | - | 9 - 10 | 1151 | 97 | |
BC032622.1 | - | 9 - 10 | 1452 | 97 | |
BC078166.1 | - | 8 - 9 | 1111 | 97 | |
BC106017.1 | - | 7 - 8 | 962 | 97 | |
BC137350.1 | - | 17 - 18 | 6508 | 97 | |
BX537849.1 | - | 18 - 19 | 6559 | 97 | |
BX538166.1 | - | 18 - 19 | 6577 | 97 | |
CR749249.1 | - | 9 - 10 | 5055 | 97 | |
KU178559.1 | - | 6 - 7 | 844 | 97 | |
KU178560.1 | - | 6 - 7 | 844 | 97 |