Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SDCCAG3
Entrez Gene ID
10807
Gene Name
serologically defined colon cancer antigen 3
Gene Aliases
NY-CO-3
Chromosome Location
Chr.9: 136401922 - 136410602 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039707.1 NP_001034796.1 6 - 7 1099 110
NM_001039708.1 NP_001034797.1 4 - 5 880 110
NM_006643.3 NP_006634.3 5 - 6 1030 110
XM_005266050.3 XP_005266107.1 6 - 7 1053 110
XM_005266051.3 XP_005266108.1 5 - 6 944 110
XM_011518157.2 XP_011516459.1 5 - 6 883 110
XM_017014218.1 XP_016869707.1 5 - 6 1200 110
XM_017014219.1 XP_016869708.1 5 - 6 1200 110
GenBank mRNA AF039688.1 - 2 - 3 167 110
AK001296.1 - 4 - 5 659 110
AY349357.1 - 5 - 6 720 110
BC014515.1 - 5 - 6 897 110

View Full Product Details