The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199420.3 | NP_955452.3 | 11 - 12 | 1945 | 101 |
XM_011512343.2 | XP_011510645.1 | 11 - 12 | 1933 | 101 | |
XM_011512347.2 | XP_011510649.1 | 11 - 12 | 1933 | 101 | |
XM_011512348.2 | XP_011510650.1 | 10 - 11 | 1802 | 101 | |
XM_011512352.2 | XP_011510654.1 | 11 - 12 | 1933 | 101 | |
XM_017005565.1 | XP_016861054.1 | 5 - 6 | 797 | 101 | |
GenBank mRNA | AB209867.1 | - | 11 - 12 | 1872 | 101 |
AY032677.1 | - | 12 - 13 | 2218 | 101 | |
AY338826.2 | - | 11 - 12 | 1945 | 101 | |
BC034254.1 | - | 2 - 3 | 312 | 101 | |
CR936627.1 | - | 10 - 11 | 1791 | 101 |