The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321309.1 | NP_001308238.1 | 8 - 9 | 844 | 73 |
NM_053025.3 | NP_444253.3 | 9 - 10 | 1052 | 73 | |
NM_053026.3 | NP_444254.3 | 9 - 10 | 1052 | 73 | |
NM_053027.3 | NP_444255.3 | 9 - 10 | 1052 | 73 | |
NM_053028.3 | NP_444256.3 | 9 - 10 | 1052 | 73 | |
XM_011512860.2 | XP_011511162.1 | 6 - 7 | 773 | 73 | |
XM_011512861.2 | XP_011511163.1 | 6 - 7 | 773 | 73 | |
GenBank mRNA | AF069601.2 | - | 7 - 8 | 889 | 73 |
AF069602.1 | - | 7 - 8 | 889 | 73 | |
AF069603.1 | - | 7 - 8 | 889 | 73 | |
AY339601.1 | - | 6 - 7 | 770 | 73 | |
AY424269.1 | - | 6 - 7 | 772 | 73 | |
AY424270.1 | - | 6 - 7 | 772 | 73 | |
U48959.2 | - | 7 - 8 | 889 | 73 |