Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SEMA5B
Entrez Gene ID
54437
Gene Name
semaphorin 5B
Gene Aliases
SEMAG, SemG
Chromosome Location
Chr.3: 122909193 - 123028605 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031702.3 NP_001026872.2 11 - 12 1866 60
NM_001256346.1 NP_001243275.1 11 - 12 2006 60
NM_001256347.1 NP_001243276.1 11 - 12 1652 60
NM_001256348.1 NP_001243277.1 10 - 11 1704 60
NR_046079.1 11 - 12 1676 60
XM_017006638.1 XP_016862127.1 10 - 11 1916 60
XM_017006639.1 XP_016862128.1 10 - 11 1658 60
GenBank mRNA AB040878.1 - 11 - 12 1632 60
AK091481.1 - 11 - 12 2006 60
AK291407.1 - 11 - 12 1783 60
AK294528.1 - 10 - 11 1602 60
AK295619.1 - 11 - 12 1652 60
AK309834.1 - 10 - 11 1583 60
AK309848.1 - 10 - 11 1658 60
AY358124.1 - 11 - 12 1676 60
BC077726.1 - 11 - 12 1549 60

View Full Product Details