The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031702.3 | NP_001026872.2 | 14 - 15 | 2369 | 70 |
NM_001256346.1 | NP_001243275.1 | 14 - 15 | 2509 | 70 | |
NM_001256347.1 | NP_001243276.1 | 14 - 15 | 2155 | 70 | |
NM_001256348.1 | NP_001243277.1 | 13 - 14 | 2207 | 70 | |
NR_046079.1 | 14 - 15 | 2179 | 70 | ||
XM_017006638.1 | XP_016862127.1 | 13 - 14 | 2419 | 70 | |
XM_017006639.1 | XP_016862128.1 | 13 - 14 | 2161 | 70 | |
GenBank mRNA | AB040878.1 | - | 14 - 15 | 2135 | 70 |
AK091481.1 | - | 14 - 15 | 2509 | 70 | |
AK291407.1 | - | 14 - 15 | 2286 | 70 | |
AK294528.1 | - | 13 - 14 | 2105 | 70 | |
AK295619.1 | - | 14 - 15 | 2155 | 70 | |
AK309848.1 | - | 13 - 14 | 2161 | 70 | |
AY358124.1 | - | 14 - 15 | 2179 | 70 | |
BC077726.1 | - | 14 - 15 | 2052 | 70 |