Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SEMA5B
Entrez Gene ID
54437
Gene Name
semaphorin 5B
Gene Aliases
SEMAG, SemG
Chromosome Location
Chr.3: 122909193 - 123028605 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031702.3 NP_001026872.2 16 - 17 2665 86
NM_001256346.1 NP_001243275.1 16 - 17 2805 86
NM_001256347.1 NP_001243276.1 16 - 17 2451 86
NM_001256348.1 NP_001243277.1 15 - 16 2503 86
XM_017006638.1 XP_016862127.1 15 - 16 2715 86
XM_017006639.1 XP_016862128.1 15 - 16 2457 86
GenBank mRNA AB040878.1 - 16 - 17 2431 86
AK091481.1 - 16 - 17 2805 86
AK291407.1 - 16 - 17 2582 86
AK294528.1 - 15 - 16 2401 86
AK295619.1 - 16 - 17 2451 86
AK309848.1 - 15 - 16 2457 86
BC077726.1 - 16 - 17 2348 86

View Full Product Details