Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SEMA5B
Entrez Gene ID
54437
Gene Name
semaphorin 5B
Gene Aliases
SEMAG, SemG
Chromosome Location
Chr.3: 122909193 - 123028605 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031702.3 NP_001026872.2 18 - 19 3113 90
NM_001256346.1 NP_001243275.1 18 - 19 3253 90
NM_001256347.1 NP_001243276.1 18 - 19 2899 90
NR_046079.1 18 - 19 2920 90
XM_017006638.1 XP_016862127.1 17 - 18 3163 90
GenBank mRNA AB040878.1 - 18 - 19 2879 90
AK091481.1 - 18 - 19 3253 90
AK291407.1 - 18 - 19 3030 90
AK295619.1 - 18 - 19 2899 90
AY358124.1 - 18 - 19 2920 90
BC077726.1 - 18 - 19 2796 90

View Full Product Details