Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SEMA5B
Entrez Gene ID
54437
Gene Name
semaphorin 5B
Gene Aliases
SEMAG, SemG
Chromosome Location
Chr.3: 122909193 - 123028605 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031702.3 NP_001026872.2 7 - 8 1024 90
NM_001256346.1 NP_001243275.1 7 - 8 1164 90
NM_001256347.1 NP_001243276.1 7 - 8 810 90
NM_001256348.1 NP_001243277.1 6 - 7 862 90
NR_046079.1 7 - 8 834 90
XM_017006638.1 XP_016862127.1 6 - 7 1074 90
XM_017006639.1 XP_016862128.1 6 - 7 816 90
GenBank mRNA AB040878.1 - 7 - 8 790 90
AK091481.1 - 7 - 8 1164 90
AK291407.1 - 7 - 8 941 90
AK294528.1 - 6 - 7 760 90
AK295619.1 - 7 - 8 810 90
AK309834.1 - 5 - 6 648 90
AK309848.1 - 6 - 7 816 90
AY358124.1 - 7 - 8 834 90
BC077726.1 - 7 - 8 707 90

View Full Product Details