The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001122770.1 | NP_001116242.1 | 4 - 5 | 1293 | 64 |
XM_005245546.2 | XP_005245603.1 | 4 - 5 | 1348 | 64 | |
XM_006711578.3 | XP_006711641.1 | 3 - 4 | 1357 | 64 | |
XM_017002556.1 | XP_016858045.1 | 3 - 4 | 1358 | 64 | |
GenBank mRNA | AF318371.1 | - | 3 - 4 | 1211 | 64 |
AK302696.1 | - | 4 - 5 | 1265 | 64 |