The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002597.4 | NP_002588.3 | 1 - 2 | 43 | 116 |
GenBank mRNA | EL945964.1 | - | 1 - 2 | 47 | 116 |