Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EDEM3
Entrez Gene ID
80267
Gene Name
ER degradation enhancing alpha-mannosidase like protein 3
Gene Aliases
C1orf22
Chromosome Location
Chr.1: 184690491 - 184754907 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001319960.1 NP_001306889.1 10 - 11 1339 131
NM_025191.3 NP_079467.3 10 - 11 1339 131
NR_135118.1 10 - 11 1339 131
XM_005245499.1 XP_005245556.1 10 - 11 1320 131
XM_011510012.1 XP_011508314.1 9 - 10 1259 131
XM_011510013.2 XP_011508315.1 10 - 11 1320 131
XM_011510014.2 XP_011508316.1 10 - 11 1320 131
XM_017002397.1 XP_016857886.1 9 - 10 1259 131
XM_017002398.1 XP_016857887.1 9 - 10 1259 131
GenBank mRNA AF288393.1 - 10 - 11 1002 131
AK225171.1 - 10 - 11 1282 131
AK315118.1 - 10 - 11 1283 131
BC144149.1 - 10 - 11 999 131
BC144150.1 - 10 - 11 999 131

View Full Product Details