The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319960.1 | NP_001306889.1 | 11 - 12 | 1422 | 127 |
NM_025191.3 | NP_079467.3 | 11 - 12 | 1422 | 127 | |
NR_135118.1 | 11 - 12 | 1422 | 127 | ||
XM_005245499.1 | XP_005245556.1 | 11 - 12 | 1403 | 127 | |
XM_011510012.1 | XP_011508314.1 | 10 - 11 | 1342 | 127 | |
XM_011510013.2 | XP_011508315.1 | 11 - 12 | 1403 | 127 | |
XM_011510014.2 | XP_011508316.1 | 11 - 12 | 1403 | 127 | |
XM_017002397.1 | XP_016857886.1 | 10 - 11 | 1342 | 127 | |
XM_017002398.1 | XP_016857887.1 | 10 - 11 | 1342 | 127 | |
GenBank mRNA | AF288393.1 | - | 11 - 12 | 1085 | 127 |
AK225171.1 | - | 11 - 12 | 1365 | 127 | |
AK315118.1 | - | 11 - 12 | 1366 | 127 | |
BC144149.1 | - | 11 - 12 | 1082 | 127 | |
BC144150.1 | - | 11 - 12 | 1082 | 127 |