The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001201550.2 | NP_001188479.1 | 8 - 9 | 1491 | 77 |
NM_001201551.1 | NP_001188480.1 | 8 - 9 | 1488 | 77 | |
NM_006684.4 | NP_006675.2 | 4 - 5 | 750 | 77 | |
XM_006711129.3 | XP_006711192.1 | 8 - 9 | 1430 | 77 | |
XM_017000111.1 | XP_016855600.1 | 8 - 9 | 1454 | 77 | |
XM_017000114.1 | XP_016855603.1 | 5 - 6 | 882 | 77 | |
GenBank mRNA | AJ640130.2 | - | 8 - 9 | 1420 | 77 |
AK292752.1 | - | 8 - 9 | 1488 | 77 | |
BC074957.2 | - | 5 - 6 | 661 | 77 |