The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001261408.1 | NP_001248337.1 | 8 - 9 | 1317 | 58 |
NM_001261409.1 | NP_001248338.1 | 6 - 7 | 1303 | 58 | |
NM_001261410.1 | NP_001248339.1 | 4 - 5 | 970 | 58 | |
NM_001308251.1 | NP_001295180.1 | 2 - 3 | 290 | 58 | |
NM_003567.3 | NP_003558.1 | 6 - 7 | 1280 | 58 | |
XM_011542251.2 | XP_011540553.1 | 5 - 6 | 919 | 58 | |
XM_017002480.1 | XP_016857969.1 | 8 - 9 | 1693 | 58 | |
GenBank mRNA | AF124250.1 | - | 6 - 7 | 1272 | 58 |
AK027785.1 | - | 2 - 3 | 538 | 58 | |
AK055442.1 | - | 6 - 7 | 1303 | 58 | |
AK301570.1 | - | 2 - 3 | 263 | 58 | |
AK310970.1 | - | 2 - 3 | 290 | 58 | |
AL833121.1 | - | 4 - 5 | 923 | 58 | |
AY358996.1 | - | 2 - 3 | 210 | 58 | |
BC028477.1 | - | 8 - 9 | 1346 | 58 | |
BC039895.1 | - | 6 - 7 | 1173 | 58 | |
U92715.1 | - | 6 - 7 | 1088 | 58 |