Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BCAR3
Entrez Gene ID
8412
Gene Name
breast cancer anti-estrogen resistance 3
Gene Aliases
NSP2, SH2D3B
Chromosome Location
Chr.1: 93561741 - 93847150 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261408.1 NP_001248337.1 12 - 13 2379 90
NM_001261409.1 NP_001248338.1 10 - 11 2365 90
NM_001261410.1 NP_001248339.1 8 - 9 2032 90
NM_001308251.1 NP_001295180.1 6 - 7 1352 90
NM_003567.3 NP_003558.1 10 - 11 2342 90
XM_011542251.2 XP_011540553.1 9 - 10 1981 90
XM_017002480.1 XP_016857969.1 12 - 13 2755 90
GenBank mRNA AF124250.1 - 10 - 11 2334 90
AK027785.1 - 6 - 7 1600 90
AK055442.1 - 10 - 11 2365 90
AK301570.1 - 6 - 7 1325 90
AL833121.1 - 8 - 9 1985 90
AY358996.1 - 6 - 7 1272 90
BC028477.1 - 12 - 13 2408 90
BC039895.1 - 10 - 11 2235 90
U92715.1 - 10 - 11 2150 90

View Full Product Details