Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAD51D
Entrez Gene ID
5892
Gene Name
RAD51 paralog D
Gene Aliases
BROVCA4, R51H3, RAD51L3, TRAD
Chromosome Location
Chr.17: 35099792 - 35119869 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB020412.1 - 1 - 2 89 88
AK302450.1 - 6 - 7 729 88

View Full Product Details