Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OFCC1
Entrez Gene ID
266553
Gene Name
orofacial cleft 1 candidate 1
Gene Aliases
MRDS1
Chromosome Location
Chr.6: 9775808 - 9842630 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF520800.1 - 2 - 3 96 67
AF520808.1 - 1 - 2 56 67
AF548113.1 - 2 - 3 242 67
AY309094.1 - 3 - 4 71
BC113541.1 - 1 - 2 35 67
BC113543.1 - 1 - 2 35 67
BC143837.1 - 1 - 2 35 67

Gene Symbol
LOC107983965
Entrez Gene ID
107983965
Gene Name
orofacial cleft 1 candidate gene 1 protein-like
Gene Aliases
-
Chromosome Location
Chr.6: 9850416 - 10211608 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017011612.1 XP_016867101.1 3 - 4 393 67

View Full Product Details