Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AKNA
Entrez Gene ID
80709
Gene Name
AT-hook transcription factor
Gene Aliases
-
Chromosome Location
Chr.9: 114334149 - 114398498 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005252244.2 XP_005252301.1 2 - 3 168 76
XM_017015172.1 XP_016870661.1 2 - 3 153 76

View Full Product Details