The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002349.3 | NP_002340.2 | 2 - 3 | 534 | 79 |
GenBank mRNA | AF011333.1 | - | 2 - 3 | 513 | 79 |
AF064827.1 | - | 2 - 3 | 460 | 79 | |
AK310678.1 | - | 2 - 3 | 520 | 79 | |
DB150194.1 | - | 2 - 3 | 520 | 79 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198759.1 | NP_001185688.1 | 2 - 3 | 534 | 79 |
NM_001198760.1 | NP_001185689.1 | 2 - 3 | 534 | 79 | |
GenBank mRNA | AY184222.1 | - | 2 - 3 | 460 | 79 |
AY314006.1 | - | 2 - 3 | 460 | 79 |