Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COL7A1
Entrez Gene ID
1294
Gene Name
collagen type VII alpha 1
Gene Aliases
EBD1, EBDCT, EBR1, NDNC8
Chromosome Location
Chr.3: 48564073 - 48595302 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000094.3 NP_000085.1 117 - 118 8825 114
XM_011533337.1 XP_011531639.1 118 - 119 8860 114
XM_017005688.1 XP_016861177.1 117 - 118 8800 114
GenBank mRNA AB209645.1 - 81 - 82 5337 114
L02870.1 - 117 - 118 8937 114
L06862.1 - 24 - 25 1642 114

View Full Product Details