The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303494.1 | NP_001290423.1 | 3 - 4 | 326 | 61 |
NM_014292.4 | NP_055107.3 | 3 - 4 | 326 | 61 | |
GenBank mRNA | AK094829.1 | - | 3 - 4 | 241 | 61 |
AK293035.1 | - | 3 - 4 | 208 | 61 | |
BC012111.1 | - | 3 - 4 | 205 | 61 | |
BC064900.1 | - | 3 - 4 | 193 | 61 | |
HY128779.1 | - | 3 - 4 | 326 | 61 | |
HY151719.1 | - | 3 - 4 | 290 | 61 |