The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321153.1 | NP_001308082.1 | 19 - 20 | 2885 | 63 |
NM_007335.3 | NP_031361.2 | 19 - 20 | 2885 | 63 | |
NM_007337.3 | NP_031363.2 | 19 - 20 | 2885 | 63 | |
XM_006713438.3 | XP_006713501.1 | 19 - 20 | 3494 | 63 | |
XM_006713439.3 | XP_006713502.1 | 19 - 20 | 3493 | 63 | |
XM_006713440.3 | XP_006713503.1 | 19 - 20 | 3494 | 63 | |
XM_011534317.2 | XP_011532619.1 | 19 - 20 | 3493 | 63 | |
XM_011534318.2 | XP_011532620.1 | 18 - 19 | 3320 | 63 | |
GenBank mRNA | AB020522.1 | - | 19 - 20 | 2885 | 63 |
AB425198.1 | - | 19 - 20 | 2857 | 63 | |
AL137706.1 | - | 7 - 8 | 1872 | 63 | |
BC171799.1 | - | 19 - 20 | 2885 | 63 |