The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321153.1 | NP_001308082.1 | 27 - 28 | 3883 | 87 |
NM_007335.3 | NP_031361.2 | 27 - 28 | 3874 | 87 | |
NM_007337.3 | NP_031363.2 | 27 - 28 | 3874 | 87 | |
XM_006713438.3 | XP_006713501.1 | 27 - 28 | 4492 | 87 | |
XM_006713439.3 | XP_006713502.1 | 27 - 28 | 4491 | 87 | |
XM_006713440.3 | XP_006713503.1 | 27 - 28 | 4492 | 87 | |
XM_011534317.2 | XP_011532619.1 | 27 - 28 | 4491 | 87 | |
XM_011534318.2 | XP_011532620.1 | 26 - 27 | 4318 | 87 | |
GenBank mRNA | AB020522.1 | - | 27 - 28 | 3874 | 87 |
AB425198.1 | - | 27 - 28 | 3855 | 87 | |
AL137706.1 | - | 15 - 16 | 2756 | 87 | |
BC171799.1 | - | 27 - 28 | 3874 | 87 |