The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321153.1 | NP_001308082.1 | 9 - 10 | 1590 | 88 |
NM_007335.3 | NP_031361.2 | 9 - 10 | 1590 | 88 | |
NM_007337.3 | NP_031363.2 | 9 - 10 | 1590 | 88 | |
XM_006713438.3 | XP_006713501.1 | 9 - 10 | 2199 | 88 | |
XM_006713439.3 | XP_006713502.1 | 9 - 10 | 2198 | 88 | |
XM_006713440.3 | XP_006713503.1 | 9 - 10 | 2199 | 88 | |
XM_011534317.2 | XP_011532619.1 | 9 - 10 | 2198 | 88 | |
XM_011534318.2 | XP_011532620.1 | 8 - 9 | 2025 | 88 | |
GenBank mRNA | AB020522.1 | - | 9 - 10 | 1590 | 88 |
AB425198.1 | - | 9 - 10 | 1562 | 88 | |
BC129824.1 | - | 9 - 10 | 1590 | 88 | |
BC129825.1 | - | 9 - 10 | 1590 | 88 | |
BC171799.1 | - | 9 - 10 | 1590 | 88 |