The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042478.1 | NP_001035943.1 | 2 - 3 | 1207 | 94 |
NM_018836.3 | NP_061324.1 | 2 - 3 | 1207 | 94 | |
XM_011541786.2 | XP_011540088.1 | 2 - 3 | 1477 | 94 | |
XM_011541787.2 | XP_011540089.1 | 2 - 3 | 1363 | 94 | |
GenBank mRNA | AF175409.1 | - | 2 - 3 | 1519 | 94 |
AL049672.1 | - | 1 - 2 | 292 | 94 | |
AL049673.1 | - | 1 - 2 | 292 | 94 | |
AY282806.1 | - | 2 - 3 | 1207 | 94 |