Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AJAP1
Entrez Gene ID
55966
Gene Name
adherens junctions associated protein 1
Gene Aliases
MOT8, SHREW-1, SHREW1
Chromosome Location
Chr.1: 4654775 - 4792518 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042478.1 NP_001035943.1 3 - 4 1301 70
NM_018836.3 NP_061324.1 3 - 4 1301 70
XM_011541786.2 XP_011540088.1 3 - 4 1571 70
XM_011541787.2 XP_011540089.1 3 - 4 1457 70
GenBank mRNA AF175409.1 - 3 - 4 1613 70
AL049672.1 - 2 - 3 386 70
AL049673.1 - 2 - 3 386 70
AY282806.1 - 3 - 4 1301 70

View Full Product Details