The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000537.3 | NP_000528.1 | 2 - 3 | 292 | 63 |
GenBank mRNA | BC033474.1 | - | 2 - 3 | 274 | 63 |
BC047752.1 | - | 2 - 3 | 277 | 63 | |
BP321283.1 | - | 2 - 3 | 292 | 63 | |
CR536498.1 | - | 2 - 3 | 248 | 63 |