Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARMCX2
Entrez Gene ID
9823
Gene Name
armadillo repeat containing, X-linked 2
Gene Aliases
ALEX2, GASP9
Chromosome Location
Chr.X: 101655281 - 101659891 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282231.1 NP_001269160.1 4 - 5 343 93
NM_177949.3 NP_808818.1 4 - 5 355 93
XM_005278109.1 XP_005278166.1 4 - 5 353 93
XM_005278113.1 XP_005278170.1 4 - 5 330 93
XM_005278116.1 XP_005278173.1 4 - 5 341 93
XM_017029989.1 XP_016885478.1 4 - 5 347 93
GenBank mRNA AB011084.1 - 4 - 5 301 93
AK291342.1 - 4 - 5 314 93
AL709063.1 - 4 - 5 314 93
BG548388.1 - 4 - 5 356 93
BX648494.1 - 4 - 5 302 93
CN391032.1 - 4 - 5 273 93

View Full Product Details