The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020949.2 | NP_066000.2 | 1 - 2 | 164 | 60 |
GenBank mRNA | AK122655.1 | - | 1 - 2 | 159 | 60 |
DA475001.1 | - | 1 - 2 | 164 | 60 |