The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291970.1 | NP_001278899.1 | 1 - 2 | 295 | 67 |
NM_006502.2 | NP_006493.1 | 1 - 2 | 295 | 67 | |
XM_011514698.2 | XP_011513000.1 | 1 - 2 | 287 | 67 | |
GenBank mRNA | AB024313.1 | - | 1 - 2 | 228 | 67 |
AF158185.1 | - | 1 - 2 | 156 | 67 | |
AK023893.1 | - | 1 - 2 | 286 | 67 | |
AK294430.1 | - | 1 - 2 | 275 | 67 | |
BC015742.1 | - | 1 - 2 | 139 | 67 | |
DC416114.1 | - | 1 - 2 | 295 | 67 |