The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291969.1 | NP_001278898.1 | 6 - 7 | 957 | 84 |
NM_001291970.1 | NP_001278899.1 | 8 - 9 | 1316 | 84 | |
NM_006502.2 | NP_006493.1 | 8 - 9 | 1316 | 84 | |
XM_005249186.3 | XP_005249243.1 | 7 - 8 | 1169 | 84 | |
XM_011514698.2 | XP_011513000.1 | 7 - 8 | 1090 | 84 | |
GenBank mRNA | AB024313.1 | - | 8 - 9 | 1249 | 84 |
AF158185.1 | - | 8 - 9 | 1177 | 84 | |
AJ420484.1 | - | 5 - 6 | 633 | 84 | |
AK023893.1 | - | 8 - 9 | 1307 | 84 | |
AK025631.1 | - | - | 644 | 84 | |
AK122807.1 | - | 5 - 6 | 1224 | 84 | |
AK307660.1 | - | 6 - 7 | 942 | 84 | |
AK316151.1 | - | 7 - 8 | 1131 | 84 | |
BC015742.1 | - | 8 - 9 | 1160 | 84 |