The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322854.1 | NP_001309783.1 | 7 - 8 | 1179 | 64 |
NM_001322855.1 | NP_001309784.1 | 8 - 9 | 1321 | 64 | |
NM_001322856.1 | NP_001309785.1 | 8 - 9 | 1278 | 64 | |
NM_001322857.1 | NP_001309786.1 | 7 - 8 | 1099 | 64 | |
NM_001322858.1 | NP_001309787.1 | 8 - 9 | 1196 | 64 | |
NM_001322859.1 | NP_001309788.1 | 5 - 6 | 805 | 64 | |
NM_001322860.1 | NP_001309789.1 | 4 - 5 | 745 | 64 | |
NM_001322861.1 | NP_001309790.1 | 4 - 5 | 455 | 64 | |
NM_001322862.1 | NP_001309791.1 | 8 - 9 | 1156 | 64 | |
NM_001322863.1 | NP_001309792.1 | 7 - 8 | 1128 | 64 | |
NM_001322864.1 | NP_001309793.1 | 6 - 7 | 696 | 64 | |
NM_001322866.1 | NP_001309795.1 | 7 - 8 | 1019 | 64 | |
NM_001322868.1 | NP_001309797.1 | 7 - 8 | 1046 | 64 | |
NM_020448.4 | NP_065181.1 | 7 - 8 | 1041 | 64 | |
XM_011541808.2 | XP_011540110.1 | 6 - 7 | 956 | 64 | |
XM_017001868.1 | XP_016857357.1 | 6 - 7 | 691 | 64 | |
XM_017001869.1 | XP_016857358.1 | 7 - 8 | 1026 | 64 | |
XM_017001870.1 | XP_016857359.1 | 8 - 9 | 1291 | 64 | |
GenBank mRNA | AK312433.1 | - | 7 - 8 | 995 | 64 |
BC063583.1 | - | 7 - 8 | 971 | 64 | |
BX640883.1 | - | 8 - 9 | 1103 | 64 | |
U90918.1 | - | 7 - 8 | 932 | 64 |