The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172.3 | NP_001163.1 | 3 - 4 | 473 | 62 |
GenBank mRNA | AK312484.1 | - | 3 - 4 | 389 | 62 |
AY074489.1 | - | 3 - 4 | 418 | 62 | |
BC001350.1 | - | 3 - 4 | 374 | 62 | |
BC008464.1 | - | 3 - 4 | 415 | 62 | |
BC029050.1 | - | 3 - 4 | 473 | 62 | |
CR536550.1 | - | 3 - 4 | 357 | 62 | |
D86724.1 | - | 3 - 4 | 377 | 62 | |
U75667.1 | - | 3 - 4 | 395 | 62 | |
U82256.1 | - | 3 - 4 | 373 | 62 |