The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006226.3 | NP_006217.3 | 2 - 3 | 3108 | 87 |
XM_005246643.3 | XP_005246700.1 | 2 - 3 | 2535 | 87 | |
XM_005246644.3 | XP_005246701.1 | 2 - 3 | 2488 | 87 | |
XM_011511351.2 | XP_011509653.1 | 3 - 4 | 3165 | 87 | |
XM_017004339.1 | XP_016859828.1 | 2 - 3 | 2582 | 87 | |
XM_017004340.1 | XP_016859829.1 | 2 - 3 | 3329 | 87 | |
GenBank mRNA | AK127514.1 | - | 3 - 4 | 3165 | 87 |
AK302673.1 | - | 2 - 3 | 2535 | 87 | |
BC101531.1 | - | 1 - 2 | 2452 | 87 | |
BC111985.1 | - | 1 - 2 | 2452 | 87 | |
BX537442.1 | - | 1 - 2 | 490 | 87 | |
D42108.1 | - | 2 - 3 | 2619 | 87 |