Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NUP37
Entrez Gene ID
79023
Gene Name
nucleoporin 37
Gene Aliases
p37
Chromosome Location
Chr.12: 102074189 - 102120139 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024057.3 NP_076962.2 7 - 8 82
GenBank mRNA AF514994.1 - 6 - 7 82
AK026271.1 - 6 - 7 82
BC000861.2 - 6 - 7 82
BM995521.1 - 3 - 4 444 82

View Full Product Details