Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HNRNPU
Entrez Gene ID
3192
Gene Name
heterogeneous nuclear ribonucleoprotein U
Gene Aliases
HNRNPU-AS1, HNRPU, SAF-A, SAFA, U21.1, hnRNP U
Chromosome Location
Chr.1: 244842123 - 244864720 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004501.3 NP_004492.2 4 - 5 1183 108
NM_031844.2 NP_114032.2 4 - 5 1240 108
XM_017001115.1 XP_016856604.1 4 - 5 1435 108
XM_017001116.1 XP_016856605.1 4 - 5 1240 108
XM_017001117.1 XP_016856606.1 4 - 5 1229 108
GenBank mRNA AF068846.1 - 4 - 5 1236 108
AF461014.1 - 1 - 2 761 108
AK126868.1 - 5 - 6 981 108
AK130669.1 - 3 - 4 289 108
AK297119.1 - 5 - 6 1074 108
AK307661.1 - 4 - 5 707 108
BC003367.1 - 4 - 5 1114 108
BC003621.2 - 4 - 5 1116 108
BC007950.2 - 4 - 5 664 108
BC015782.1 - 4 - 5 717 108
BC024767.1 - 4 - 5 1116 108
BC034925.1 - 1 - 2 107 108
X65488.1 - 4 - 5 1006 108

View Full Product Details