The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000480.2 | NP_000471.1 | 3 - 4 | 797 | 58 |
NM_001025389.1 | NP_001020560.1 | 3 - 4 | 760 | 58 | |
NM_001025390.1 | NP_001020561.1 | 3 - 4 | 899 | 58 | |
NM_001172430.1 | NP_001165901.1 | 3 - 4 | 724 | 58 | |
GenBank mRNA | AK289998.1 | - | 3 - 4 | 531 | 58 |
AK302970.1 | - | 3 - 4 | 715 | 58 | |
AK308181.1 | - | 3 - 4 | 567 | 58 | |
AK311436.1 | - | 4 - 5 | 604 | 58 | |
AK311607.1 | - | 3 - 4 | 820 | 58 | |
AK311711.1 | - | 3 - 4 | 784 | 58 | |
AK313039.1 | - | 3 - 4 | 531 | 58 | |
BC126118.1 | - | 3 - 4 | 792 | 58 | |
D12775.1 | - | 3 - 4 | 616 | 58 | |
M84720.1 | - | 3 - 4 | 631 | 58 | |
M84721.1 | - | 3 - 4 | 559 | 58 | |
M84722.1 | - | 3 - 4 | 683 | 58 |