The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138785.3 | NP_620140.1 | 3 - 4 | 298 | 140 |
GenBank mRNA | AK289402.1 | - | 3 - 4 | 285 | 140 |
AK315729.1 | - | 3 - 4 | 295 | 140 | |
AY358952.1 | - | 3 - 4 | 416 | 140 | |
BC014320.1 | - | 3 - 4 | 302 | 140 |